Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031037.2 | NP_112299.1 | 5 | 470 | ||
GenBank mRNA | AF277892.1 | - | 4 | 251 | ||
AF397193.1 | - | 1 | 50 | |||
BC065579.1 | - | 5 | 470 | 5598923 | ||
DQ120485.1 | - | 4 | 235 | |||
DQ120486.1 | - | 4 | 235 | |||
U34959.1 | - | 4 | 242 |