Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190162.1 | NP_001177091.1 | 4 | 432 | ||
NM_001190163.1 | NP_001177092.1 | 4 | 420 | |||
NM_031511.2 | NP_113699.2 | 4 | 1407 | |||
XM_006230664.2 | XP_006230726.1 | 4 | 1209 | |||
XM_006230665.2 | XP_006230727.1 | 4 | 412 | |||
XM_008760074.1 | XP_008758296.1 | 4 | 1230 | |||
XM_008760075.1 | XP_008758297.1 | 5 | 745 | |||
GenBank mRNA | JQ408382.1 | - | 1345 | |||
M30273.1 | - | 2 | 161 | |||
M38688.1 | - | 3 | 266 | |||
X00911.1 | - | 4 | 1407 | |||
X13101.1 | - | 4 | 444 | |||
X14833.1 | - | 4 | 720 | |||
X14834.1 | - | 4 | 1475 | |||
X16703.1 | - | 1 | 58 |