Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ramp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031645.1 | NP_113833.1 | 2 | 206 | ||
XM_006245473.2 | XP_006245535.1 | 2 | 1070 | |||
XM_006245474.2 | XP_006245536.1 | 2 | 282 | |||
GenBank mRNA | AB030942.1 | - | 2 | 182 | ||
AB042887.1 | - | 2 | 206 | |||
AF181550.1 | - | 2 | 183 |