Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138907.2 | NP_620262.2 | 1 | 590 | ||
XM_006240320.2 | XP_006240382.1 | 1 | 674 | |||
GenBank mRNA | AB010429.1 | - | 1 | 600 | ||
BC072540.1 | - | 1 | 590 | 7104319 | ||
Y09333.1 | - | 1 | 531 |