Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139083.2 | NP_620783.1 | 4 | 269 | ||
GenBank mRNA | BC099836.1 | - | 3 | 210 | ||
BC126080.1 | - | 3 | 206 | |||
BC157821.1 | - | 3 | 199 | |||
FQ211334.1 | - | 4 | 223 | |||
FQ217332.1 | - | 4 | 269 | |||
FQ217674.1 | - | 4 | 222 | |||
FQ220278.1 | - | 4 | 214 | |||
FQ221387.1 | - | 4 | 218 | |||
FQ221514.1 | - | 4 | 214 | |||
FQ221865.1 | - | 4 | 220 | |||
FQ222435.1 | - | 4 | 222 | |||
FQ222632.1 | - | 4 | 222 | |||
FQ222800.1 | - | 4 | 214 | |||
FQ222813.1 | - | 4 | 222 | |||
FQ223254.1 | - | 4 | 219 | |||
FQ224594.1 | - | 4 | 222 | |||
FQ228513.1 | - | 4 | 217 | |||
FQ229040.1 | - | 4 | 221 | |||
FQ229533.1 | - | 4 | 222 | |||
X82550.1 | - | 4 | 215 |