Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031037.2 | NP_112299.1 | 7 | 896 | ||
GenBank mRNA | AF277892.1 | - | 6 | 677 | ||
AF397193.1 | - | 3 | 476 | |||
BC065579.1 | - | 7 | 896 | 5598923 | ||
DQ120485.1 | - | 6 | 661 | |||
DQ120486.1 | - | 6 | 661 | |||
FQ209560.1 | - | 3 | 284 | |||
U34959.1 | - | 6 | 668 |