Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031851.2 | NP_114039.1 | 7 | 1689 | ||
GenBank mRNA | BC072518.1 | - | 7 | 1689 | 7105633 | |
FQ229103.1 | - | 7 | 1691 | |||
M61219.1 | - | 6 | 1638 |