Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053354.3 | NP_445806.3 | 2 | 186 | ||
GenBank mRNA | AB012214.1 | - | 2 | 170 | ||
AF116344.1 | - | 2 | 224 | |||
D64060.1 | - | 2 | 58 | |||
DV214494.1 | - | 2 | 152 |