Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270867.1 | NP_001257796.1 | 3 | 808 | ||
NM_053601.2 | NP_446053.1 | 3 | 862 | |||
NM_181687.2 | NP_859015.1 | 2 | 781 | |||
XM_008762390.1 | XP_008760612.1 | 3 | 852 | |||
GenBank mRNA | BC127473.1 | - | 2 | 715 | 8364201 | |
FQ213121.1 | - | 3 | 820 | |||
FQ213576.1 | - | 2 | 739 | |||
U08290.1 | - | 3 | 779 | |||
U09785.1 | - | 2 | 698 |