Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003222.3 | NP_003213.1 | 7 | 1597 | ||
GenBank mRNA | AK301572.1 | - | 7 | 1413 | ||
BC035664.1 | - | 7 | 1556 | 5723548 | ||
BC051829.1 | - | 7 | 1597 | 6148554 | ||
U85658.1 | - | 7 | 1520 | |||
X95693.1 | - | 7 | 1400 |