Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138907.2 | NP_620262.2 | 3 | 1134 | ||
XM_006240320.2 | XP_006240382.1 | 3 | 1218 | |||
GenBank mRNA | BC072540.1 | - | 3 | 1134 | 7104319 | |
Y09333.1 | - | 3 | 1075 |