Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003253.2 | NP_003244.2 | 11 | 2664 | ||
XM_005261037.1 | XP_005261094.1 | 10 | 2797 | |||
XM_005261038.2 | XP_005261095.1 | 9 | 2531 | |||
XM_005261040.2 | XP_005261097.1 | 10 | 2797 | |||
XM_011529711.1 | XP_011528013.1 | 11 | 2748 | |||
XM_011529712.1 | XP_011528014.1 | 11 | 2672 | |||
XM_011529713.1 | XP_011528015.1 | 11 | 3082 | |||
XM_017028447.1 | XP_016883936.1 | 11 | 2851 | |||
XM_017028448.1 | XP_016883937.1 | 11 | 2821 | |||
XM_017028449.1 | XP_016883938.1 | 10 | 2728 | |||
XM_017028450.1 | XP_016883939.1 | 10 | 2748 | |||
XM_017028451.1 | XP_016883940.1 | 10 | 3228 | |||
XM_017028452.1 | XP_016883941.1 | 12 | 3028 | |||
XM_017028453.1 | XP_016883942.1 | 10 | 2703 | |||
GenBank mRNA | AB209101.1 | - | 7 | 1680 | ||
AK093621.1 | - | 9 | 1521 | |||
BC117192.1 | - | 8 | 2239 | 40125743 | ||
BC117196.1 | - | 8 | 2239 | 40125747 | ||
BC143970.1 | - | 8 | 2239 | |||
U16296.1 | - | 11 | 2665 |