Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172326.1 | NP_758829.1 | 2, 3 | 337 | ||
GenBank mRNA | AY149902.1 | - | 2, 3 | 337 |