Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173124.1 | NP_775147.1 | 157 | |||
XM_006241051.2 | XP_006241113.1 | 2 | 226 | |||
XM_008765157.1 | XP_008763379.1 | 2 | 228 | |||
XM_008765158.1 | XP_008763380.1 | 2 | 228 | |||
GenBank mRNA | BC129085.1 | - | 2 | 118 | 7375658 | |
U39207.1 | - | 157 |
Gene Symbol: Cyp4f37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271352.1 | NP_001258281.1 | 1 | 84 |