Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173126.3 | NP_775149.1 | 2 | 497 | ||
XM_006254762.2 | XP_006254824.1 | 4 | 1180 | |||
XM_006254763.2 | XP_006254825.1 | 4 | 1183 | |||
XM_006254765.2 | XP_006254827.1 | 4 | 709 | |||
XM_006254767.1 | XP_006254829.1 | 2 | 183 | |||
XM_006254769.2 | XP_006254831.1 | 2 | 386 | |||
XM_008772096.1 | XP_008770318.1 | 3 | 1311 | |||
XM_008772097.1 | XP_008770319.1 | 2 | 1119 | |||
GenBank mRNA | AF313411.1 | - | 2 | 296 | ||
BC070515.1 | - | 2 | 497 | 7100997 | ||
EV780394.1 | - | 2 | 278 |