Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173135.1 | NP_775158.1 | 1 | 79 | ||
XM_008762632.1 | XP_008760854.1 | 1 | 95 | |||
GenBank mRNA | AF013598.1 | - | 1 | 79 |