Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172040.2 | NP_742037.2 | 2 | 371 | ||
XM_006243855.2 | XP_006243917.1 | 2 | 392 | |||
GenBank mRNA | AF034218.1 | - | 2 | 341 | ||
AF535141.1 | - | 1 | 279 | |||
BC062410.1 | - | 2 | 371 | 5623650 |