Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vimp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173120.2 | NP_775143.2 | 2 | 155 | ||
GenBank mRNA | AF367467.1 | - | 2 | 165 | ||
BC059111.1 | - | 2 | 159 | 6920258 | ||
FQ210095.1 | - | 2 | 147 | |||
FQ218853.1 | - | 2 | 165 | |||
FQ219944.1 | - | 2 | 147 | |||
FQ220082.1 | - | 2 | 150 | |||
FQ225575.1 | - | 2 | 165 | |||
FQ229642.1 | - | 2 | 147 |