Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryge | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173289.1 | NP_775411.1 | 3 | 316 | ||
XM_008767178.1 | XP_008765400.1 | 4 | 493 | |||
GenBank mRNA | J00716.1 | - | 274 |
Gene Symbol: Crygf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109557.1 | NP_001103027.1 | 3 | 369 |