Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003403.4 | NP_003394.1 | 5 | 1860 | ||
GenBank mRNA | BC037308.1 | - | 5 | 1492 | 5261384 | |
BC065366.1 | - | 5 | 1425 | 5815774 | ||
BX647456.1 | - | 1 | 2007 | |||
M76541.1 | - | 5 | 1454 | |||
M77698.1 | - | 5 | 1620 | |||
Z14077.1 | - | 5 | 1635 |