Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173125.1 | NP_775148.1 | 7 | 632 | ||
XM_006253610.2 | XP_006253672.2 | 7 | 823 | |||
XM_006253611.2 | XP_006253673.2 | 7 | 925 | |||
XM_006253612.2 | XP_006253674.2 | 7 | 823 | |||
XM_008771487.1 | XP_008769709.1 | 7 | 925 | |||
GenBank mRNA | AF095585.1 | - | 7 | 632 | ||
BC078693.1 | - | 7 | 624 | 7121395 |