Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173129.1 | NP_775152.1 | 2 | 412 | ||
GenBank mRNA | AF010293.1 | - | 2 | 412 |
Gene Symbol: LOC100909862 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232222.1 | XP_006232284.1 | 1 | 202 |
Gene Symbol: LOC100910467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749189.1 | XP_003749237.1 | 1 | 202 |
Gene Symbol: LOC103693054 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766107.1 | XP_008764329.1 | 1 | 202 |