Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173139.1 | NP_775162.1 | 3 | 314 | ||
GenBank mRNA | X65747.1 | - | 3 | 314 |