Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173298.2 | NP_775420.2 | 1 | 940 | ||
XM_008763052.1 | XP_008761274.1 | 3 | 1416 | |||
GenBank mRNA | AY510305.1 | - | 1 | 940 | ||
U36899.1 | - | 3 | 1409 |