Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC286960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173301.1 | NP_775423.1 | 3 | 280 | ||
XM_008762764.1 | XP_008760986.1 | 2 | 431 | |||
GenBank mRNA | X15679.1 | - | 3 | 280 |