Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175594.2 | NP_783184.1 | 1 | 302 | ||
XM_006247193.2 | XP_006247255.1 | 1 | 623 | |||
GenBank mRNA | AF541940.1 | - | 1 | 302 | ||
BC161831.1 | - | 1 | 334 | 9023972 |