Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175598.2 | NP_783188.1 | 1, 2 | 163 | ||
XM_006241180.2 | XP_006241242.1 | 1, 2 | 778 | |||
XM_006241182.2 | XP_006241244.1 | 1, 2 | 778 | |||
XM_008765234.1 | XP_008763456.1 | 1, 2 | 778 | |||
XM_008765236.1 | XP_008763458.1 | 1, 2 | 778 | |||
XM_008765237.1 | XP_008763459.1 | 1, 2 | 777 | |||
GenBank mRNA | AY177755.1 | - | 1, 2 | 163 |