Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnmb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176861.1 | NP_789831.1 | 2 | 77 | ||
GenBank mRNA | AY191836.1 | - | 2 | 77 | ||
BC081695.1 | - | 377 | 7190722 |