Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003441.2 | NP_003432.1 | 4 | 532 | ||
XM_011513562.2 | XP_011511864.1 | 3 | 1362 | |||
XM_011513563.2 | XP_011511865.1 | 2 | 1120 | |||
XM_017008591.1 | XP_016864080.1 | 4 | 1454 | |||
GenBank mRNA | BC074853.2 | - | 4 | 392 | 30915571 | |
DA385292.1 | - | 4 | 531 | |||
L15309.1 | - | 4 | 510 |
Gene Symbol: LOC441666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024380.1 | 3 | 412 |