Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282656.1 | NP_001269585.1 | 4 | 355 | ||
NM_001282657.1 | NP_001269586.1 | 3 | 262 | |||
NM_003442.5 | NP_003433.3 | 4 | 355 | |||
XM_005253121.1 | XP_005253178.1 | 4 | 303 | |||
XM_005253122.1 | XP_005253179.1 | 4 | 331 | |||
XM_005253127.1 | XP_005253184.1 | 5 | 381 | |||
XM_011520349.2 | XP_011518651.1 | 4 | 1074 | |||
XM_017018252.1 | XP_016873741.1 | 3 | 239 | |||
XM_017018253.1 | XP_016873742.1 | 3 | 239 | |||
XM_017018254.1 | XP_016873743.1 | 4 | 343 | |||
XM_017018255.1 | XP_016873744.1 | 4 | 433 | |||
XM_017018256.1 | XP_016873745.1 | 3 | 245 | |||
GenBank mRNA | AK291133.1 | - | 4 | 308 | ||
AK297214.1 | - | 3 | 207 | |||
AK313330.1 | - | 4 | 344 | |||
BC020219.1 | - | 4 | 354 | 4902668 | ||
BG825506.1 | - | 4 | 355 |