Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eddm3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178103.2 | NP_835204.1 | 1 | 117 | ||
XM_008770507.1 | XP_008768729.1 | 2 | 630 | |||
GenBank mRNA | AJ515238.1 | - | 1 | 117 |
Gene Symbol: LOC103693384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773793.1 | XP_008772015.1 | 2 | 296 |
Gene Symbol: LOC103693823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770629.1 | XP_008768851.1 | 2 | 529 |