Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181631.2 | NP_853662.1 | 22 | 2733 | ||
XM_008764477.1 | XP_008762699.1 | 22 | 4507 | |||
GenBank mRNA | AY274810.1 | - | 22 | 2733 | ||
FQ228097.1 | - | 5 | 675 |