Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003447.3 | NP_003438.1 | 2 | 913 | ||
XM_017011258.1 | XP_016866747.1 | 2 | 118 | |||
XM_017011259.1 | XP_016866748.1 | 2 | 300 | |||
XM_017011260.1 | XP_016866749.1 | 2 | 164 | |||
XM_017011262.1 | XP_016866751.1 | 2 | 118 | |||
GenBank mRNA | AK292371.1 | - | 2 | 300 | ||
AY083664.1 | - | 3 | 1169 | |||
AY366500.1 | - | 2 | 279 | |||
BC026092.1 | - | 2 | 170 | 4837021 | ||
BC037779.2 | - | 2 | 291 | 5272359 | ||
U78722.1 | - | 2 | 653 | |||
X84801.1 | - | 2 | 324 |