Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1c9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175759.2 | NP_786935.1 | 2, 3 | 245 | ||
GenBank mRNA | BC061771.1 | - | 2, 3 | 237 | 5621994 | |
M11566.1 | - | 2, 3 | 245 |