Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301779.1 | NP_001288708.1 | 4 | 825 | ||
NM_001301780.1 | NP_001288709.1 | 3 | 681 | |||
NM_001301819.1 | NP_001288748.1 | 2 | 542 | |||
NM_003455.3 | NP_003446.2 | 5 | 877 | |||
XM_005271659.1 | XP_005271716.1 | 5 | 909 | |||
XM_005271660.1 | XP_005271717.1 | 5 | 918 | |||
XM_005271661.1 | XP_005271718.1 | 4 | 789 | |||
XM_005271664.1 | XP_005271721.1 | 3 | 614 | |||
XM_006718901.2 | XP_006718964.1 | 6 | 982 | |||
XM_011542972.1 | XP_011541274.1 | 4 | 1078 | |||
XM_011542973.1 | XP_011541275.1 | 5 | 1188 | |||
XM_011542975.1 | XP_011541277.1 | 5 | 1351 | |||
XM_011542976.1 | XP_011541278.1 | 4 | 1074 | |||
XM_017018268.1 | XP_016873757.1 | 6 | 1049 | |||
GenBank mRNA | AF027218.1 | - | 2 | 455 | ||
AF027219.1 | - | 2 | 459 | |||
AF155101.1 | - | 2 | 147 | |||
AK023751.1 | - | 3 | 615 | |||
AK226047.1 | - | 4 | 824 | |||
AL162031.1 | - | 2 | 409 | |||
BC013382.2 | - | 5 | 849 | 3508386 | ||
DA770117.1 | - | 2 | 267 |