Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6pc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176077.3 | NP_788266.2 | 1, 2 | 396 | ||
GenBank mRNA | AY186240.1 | - | 1, 2 | 226 | ||
BC078689.1 | - | 1, 2 | 396 | 7134166 | ||
FQ231610.1 | - | 1, 2 | 292 |