Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178091.4 | NP_835192.1 | 1 | 214 | ||
XM_006249667.2 | XP_006249729.1 | 2 | 679 | |||
XM_006249668.2 | XP_006249730.1 | 2 | 527 | |||
XM_006249669.2 | XP_006249731.1 | 2 | 494 | |||
GenBank mRNA | AY152392.1 | - | 1 | 214 | ||
BC085682.1 | - | 2 | 352 | 7189822 |