Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178101.3 | NP_835202.2 | 2 | 482 | ||
GenBank mRNA | AJ430859.1 | - | 2 | 221 | ||
BC085683.1 | - | 2 | 262 | 7110561 | ||
CO386873.1 | - | 2 | 396 |