Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173331.2 | NP_775453.1 | 4 | 300 | ||
XM_006241777.2 | XP_006241839.1 | 5 | 716 | |||
XM_006241779.2 | XP_006241841.1 | 5 | 716 | |||
XM_008765547.1 | XP_008763769.1 | 5 | 568 | |||
GenBank mRNA | AF078798.2 | - | 4 | 416 |