Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003472.3 | NP_003463.1 | 2, 3 | 336 | ||
XM_017011273.1 | XP_016866762.1 | 2, 3 | 308 | |||
GenBank mRNA | AK298089.1 | - | 2, 3 | 248 | ||
AK312616.1 | - | 2, 3 | 265 | |||
AU131262.1 | - | 2, 3 | 336 | |||
BC035259.1 | - | 2, 3 | 308 | 5122743 | ||
X64229.1 | - | 2, 3 | 176 |