Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175766.3 | NP_786942.1 | 928 | |||
GenBank mRNA | BC089766.1 | - | 6 | 933 | 7368342 | |
U39943.1 | - | 6 | 928 | |||
U40000.1 | - | 942 | ||||
U40004.1 | - | 931 |