Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142915.1 | NP_001136387.1 | 2 | 366 | ||
NM_175869.3 | NP_787065.2 | 2 | 366 | |||
GenBank mRNA | AJ430860.1 | - | 2 | 428 | ||
AJ430861.1 | - | 2 | 428 |