Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012498.1 | NP_036630.1 | 2 | 136 | ||
GenBank mRNA | BC062034.1 | - | 2 | 121 | 5623700 | |
X05884.1 | - | 2 | 133 |