Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012564.3 | NP_036696.2 | 2 | 132 | ||
GenBank mRNA | BC078891.1 | - | 2 | 132 | 7130137 | |
FQ209578.1 | - | 2 | 142 | |||
FQ209652.1 | - | 2 | 143 | |||
FQ209886.1 | - | 2 | 141 | |||
FQ210204.1 | - | 2 | 140 | |||
FQ210734.1 | - | 2 | 141 | |||
FQ219081.1 | - | 2 | 141 | |||
FQ219502.1 | - | 2 | 139 | |||
J05148.1 | - | 2 | 108 | |||
M12450.1 | - | 2 | 88 |