Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012568.2 | NP_036700.1 | 2 | 657 | ||
XM_003752039.3 | XP_003752087.1 | 3 | 281 | |||
XM_008773152.1 | XP_008771374.1 | 2 | 650 | |||
XM_008773153.1 | XP_008771375.1 | 2 | 63 | |||
GenBank mRNA | AJ310837.1 | - | 2 | 106 | ||
X61159.1 | - | 2 | 657 |