Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109968.1 | NP_001103438.1 | 2 | 473 | ||
NM_012569.2 | NP_036701.2 | 2 | 473 | |||
XM_008767075.1 | XP_008765297.1 | 2 | 644 | |||
GenBank mRNA | M65150.1 | - | 2 | 480 |