Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012573.3 | NP_036705.3 | 2, 3 | 644 | ||
GenBank mRNA | D13211.1 | - | 1, 2 | 437 | ||
M91561.1 | - | 2, 3 | 644 |