Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012488.2 | NP_036620.2 | 3 | 383 | ||
GenBank mRNA | AY921651.1 | - | 3 | 342 | ||
BC098922.1 | - | 3 | 383 | 7458629 | ||
FQ223246.1 | - | 3 | 405 | |||
FQ228339.1 | - | 3 | 582 | |||
J02635.1 | - | 3 | 405 |
Gene Symbol: LOC100911545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749828.2 | XP_003749876.1 | 689 |