Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012490.2 | NP_036622.2 | 4 | 710 | ||
XM_006242178.2 | XP_006242240.1 | 4 | 1592 | |||
XM_008765665.1 | XP_008763887.1 | 5 | 1907 | |||
XM_008765666.1 | XP_008763888.1 | 5 | 1432 | |||
GenBank mRNA | BC097345.1 | - | 4 | 710 | 7455117 | |
X59254.1 | - | 4 | 711 |