Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012504.1 | NP_036636.1 | 2 | 329 | ||
GenBank mRNA | BC061968.1 | - | 2 | 340 | 5598014 | |
M14511.1 | - | 2 | 329 | |||
M28647.1 | - | 2 | 352 | |||
X05882.1 | - | 2 | 106 |